Learn More
ER beta/NR3A2 Antibody (PPG5/25), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP261942IR
Catalog No. NBP261942IR
Add to Cart
Description
Description
ER beta/NR3A2 Monoclonal specifically detects ER beta/NR3A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
ER beta/NR3A2
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
ESR2
Tuberculin conjugated peptide that recognizes C terminus of ER beta5 at sequence LLSHVRHARYAP
0.1 mL
Primary
Human
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
PPG5/25
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2
Mouse
Protein A purified
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators
2100
Store at 4C in the dark.
IgG1
Recommendations
Recommendations