Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ER beta/NR3A2, Mouse anti-Human, Clone: 1013506, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $298.50

Specifications

Antigen ER beta/NR3A2
Clone 1013506
Applications Immunocytochemistry
Host Species Mouse
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB71061100
View Documents
R&D Systems
MAB71061100
100 ug
Each for $298.50
Add to cart
 
MAB71061SP
View Documents
R&D Systems
MAB71061SP
25 ug
Each for $119.60
Add to cart
 
Description

Description

ER beta/NR3A2 Monoclonal antibody specifically detects ER beta/NR3A2 in Human Samples. It is validated for Immunocytochemistry.
Specifications

Specifications

ER beta/NR3A2
Immunocytochemistry
Human
Primary
1013506
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit