Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ER beta/NR3A2 Mouse anti-Human, DyLight 405, Clone: 4D2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP141193V

Catalog No. NBP141193V

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ER beta/NR3A2
4D2
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Affinity Purified
ESR2
Amino acids 1-153 of human ER expressed in E. coli.
Immunogen affinity purified
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primary
2100
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2
Mouse
IgG2b
100 ul
Store at 4C in the dark.
Monoclonal
This antibody will recognize human ER beta.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit