Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ER beta/NR3A2 Mouse anti-Human, DyLight 405, Clone: 57/3, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261943V
Description
ER beta/NR3A2 Monoclonal antibody specifically detects ER beta/NR3A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
ER beta/NR3A2 | |
57/3 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Purified | |
ESR2 | |
Recognizes ER beta2 isomer at sequence CGMKMETLLPEATMEQ | |
Protein A purified | |
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
Primary | |
2100 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2 | |
Mouse | |
IgG1 | |
0.1 ml | |
Store at 4C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title