Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ER beta/NR3A2 Mouse anti-Human, Mouse, Monkey, Rabbit, PE/Atto594, Clone: 14C8, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NB200305PEATT594
Description
ER beta/NR3A2 Monoclonal antibody specifically detects ER beta/NR3A2 in Human, Mouse, Monkey, Rabbit samples. It is validated for Flow Cytometry.Specifications
ER beta/NR3A2 | |
14C8 | |
Flow Cytometry | |
Conjugated, Purified | |
ESR2 | |
Amino acids 1-153 of human ER-beta expressed in E. coli. | |
Protein G purified | |
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
Primary | |
2100.0 | |
Human, Monkey, Mouse, Rabbit |
Flow Cytometry | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2 | |
Mouse | |
IgG2b | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Strong Erb staining in epithelial cell nuclei. Occasional weak to moderate staining is seen in surrounding stromal and endothelial cell nuclei. Sporadic light cytoplasmic staining is sometimes observed. We have also successfully detected ERb in colon and ovarian tumors using the same antibody. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title