Learn More
ER beta/NR3A2 Mouse anti-Human, Rat, Porcine, Avian, Primate, Cynomolgus Monkey, Reptile, Rhesus Macaque, Sheep, DyLight 350, Clone: PPG5/10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP261944UV
Catalog No. NBP261944UV
Add to cart
Description
Description
ER beta/NR3A2 Monoclonal antibody specifically detects ER beta/NR3A2 in Human, Rat, Porcine, Avian, Primate, Cynomolgus Monkey, Reptile, Rhesus Macaque, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
ER beta/NR3A2
PPG5/10
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
ESR2
Synthetic peptide CEDSKSKEGSQNPQSQ derived from the C terminus of human ER beta1
Protein A purified
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primary
2100
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Avian, Cynomolgus Monkey, Human, Porcine, Primate, Rat, Reptile, Rhesus Macaque, Sheep