Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERAB Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI10B4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270871AF405

Catalog No. N270871A405


Add to cart

Description

Description

ERAB Monoclonal antibody specifically detects ERAB in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

ERAB
OTI10B4
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
HSD17B10
Full length recombinant protein of human ERAB (NP_004484) produced in HEK293T cells.
Immunogen affinity purified
Cancer, Lipid and Metabolism
Primary
3028.0
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
17-beta-HSD 10, 17-beta-hydroxysteroid dehydrogenase 10, 17b-HSD10, 3-hydroxy-2-methylbutyryl-CoA dehydrogenase, ABAD, amyloid-beta peptide binding alcohol dehydrogenase, CAMR, DUPXp11.22, EC 1.1.1.178, EC 1.1.1.35, Endoplasmic reticulum-associated amyloid beta-peptide-binding protein, ERAB3-hydroxyacyl-CoA dehydrogenase type-2, HADH2AB-binding alcohol dehydrogenase, hydroxyacyl-Coenzyme A dehydrogenase, type II, hydroxyacyl-Coenzyme Adehydrogenase, type II, hydroxysteroid (17-beta) dehydrogenase 10, mental retardation, X-linked, syndromic 10, MHBD, Mitochondrial ribonuclease P protein 2,3-hydroxyacyl-CoA dehydrogenase type II, Mitochondrial RNase P protein 2, MRPP2HCD2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1, short chain dehydrogenase/reductase family 5C, member 1, short chain L-3-hydroxyacyl-CoA dehydrogenase type 2, short chain type dehydrogenase/reductase XH98G2, Short-chain type dehydrogenase/reductase XH98G2, Type II HADH, XH98G2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit