Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ERAS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536459
Description
This antibody detects endogenous protein at a molecular weight of 25 kDa. Purity is >95% by SDS-PAGE.
The human ERAS gene was initially reported as a processed pseudogene (HRasp, Ha-Ras2), with several base substitutions in coding regions, but was more recently described as potentially encoding a functional human ERAS protein. The ERAS oncogene supports the tumorigenic growth of embryonic stem cells by producing a constitutively active Ras protein. ERAS protein has less than 50% amino acid identity with H-Ras, another member of Ras family but the key domains essential for Ras function are highly conserved. The protein also contains amino-acid residues identical to those present in active mutants of Ras and causes oncogenic transformation in NIH 3T3 cells. Human ERAS gene is localized in the chromosomal region Xp11.23.
Specifications
ERAS | |
Polyclonal | |
Unconjugated | |
ERAS | |
ecat5; Embryonic stem cell-expressed Ras; ERAS; E-Ras; ES cell expressed Ras; ES cell-expressed Ras; GTPase ERas; Ha-Ras2; HRAS2; HRasp; small GTPase E-Ras; small GTPase protein E-Ras; v-Ha-ras Harvey rat sarcoma viral oncogene homolog 2; v-Ha-ras Harvey rat sarcoma viral oncogene homolog pseudogene | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3266, 353283, 679682 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide; pH 7.2 | |
Q7TN89, Q7Z444 | |
ERAS | |
Synthetic peptide corresponding to amino acids 181-233 of Human E-Ras. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction