Learn More
ErbB2 (HER-2) Mouse anti-Human, Clone: MJD2, eBioscience™ Invitrogen™
Mouse Monoclonal Antibody
Manufacturer: LIFE TECHNOLOGIES 14975737

Description
This MJD2 MAb reacts with HER-2, the product of the ErbB2 gene and a member of the human epidermal growth factor receptor (EGFR) family. HER-2 is a 185 kDa transmembrane receptor with an extracellular binding domain and an intracellular tyrosine kinase domain. Activation of HER-2 occurs following formation of homodimers or heterodimers with ligand-bound EGFR family members, HER3 or HER4. Overexpression of HER-2 results in auto-phosphorylation and increased proliferation and invasiveness and is associated with subsets of multiple cancer types such as breast, ovarian, cervical, uterine, and gastric. The use of HER-2 targeted agents has been used extensively to treat HER-2-positive breast cancer, but a recent study found that these agents can also block the proliferation of breast cancer stem cells found in HER-2-negative patients and suggests the importance of HER-2 as a target for multiple types of breast cancer. The MJD2 antibody recognizes the extracellular domain of HER-2 but at an epitope distinct from that recognized by the 4D5 (Herceptin) and SP3 monoclonal antibodies.
ErbB2 (HER-2) Monoclonal antibody specifically detects ErbB2 (HER-2) in Human samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western BlotSpecifications
ErbB2 (HER-2) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P04626 | |
Mouse | |
2 mg | |
Primary | |
Human | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
MJD2 | |
Unconjugated | |
ERBB2 | |
avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; Proto-oncogene c-ErbB-2; proto-oncogene NEU; Receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb-b2 avian erythroblastic leukemia viral oncoprotein 2; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian) | |
Affinity chromatography | |
RUO | |
2064 | |
Antibody | |
IgG1 κ |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.