Learn More
ErbB3 (HER-3) Mouse anti-Human, BV750, Clone: SGP1, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 751783
Catalog No. BDB751783
Add to cart
Description
Description
ErbB3 (HER-3) Monoclonal antibody specifically detects ErbB3 (HER-3) in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
ErbB3 (HER-3)
Flow Cytometry
0.2 mg/mL
Hu ErbB3 (HER-3) BV750 SGP1 50 μg
Mouse
IgG1, κ
RUO
Primary
ErbB3 (HER-3)
Human
SGP1
Brilliant Violet 750
BV750
Human cEerbB3 Recombinant Protein
1 x 50 μg
Refrigerate (2 to 8°C)
Monoclonal
Human