Learn More
ErbB3/Her3 Antibody (OTI3E10), DyLight 680, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270661FR
Catalog No. NBP270661FR
Add to Cart
Description
Description
ErbB3/Her3 Monoclonal specifically detects ErbB3/Her3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
Specifications
ErbB3/Her3
Monoclonal
DyLight 680
PBS with 0.05% Sodium Azide
ERBB3
Full length recombinant protein of human ErbB3/Her3 (NP_001973) produced in SF9 cell.
0.1 mL
Primary
Human, Mouse, Rat
IgG2b
Immunohistochemistry
OTI3E10
Immunohistochemistry
c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Mouse
Affinity Purified
Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors
2065.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions