Learn More
ErbB3/Her3 Mouse anti-Human, Alexa Fluor™ 594, Clone: SGP1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB1002692AF594
Catalog No. N102692A594
Add to cart
Description
Description
ErbB3/Her3 Monoclonal antibody specifically detects ErbB3/Her3 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
ErbB3/Her3
SGP1
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
ERBB3
c-erb B3 protein from transfected human kidney fibroblasts purified by wheatgerm lectin affinity chromatography
0.1 mL
Primary
2065