Learn More
ErbB3/Her3 Rabbit anti-Human, Alexa Fluor™ 750, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232253AF750
Catalog No. NP23223A70
Add to cart
Description
Description
ErbB3/Her3 Polyclonal antibody specifically detects ErbB3/Her3 in Human samples. It is validated for Western Blot.
Specifications
Specifications
ErbB3/Her3
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
ERBB3
The immunogen this antibody was made to, maps to a region between residue 1292 to 1342 of human V-Erb-B2 Erythroblastic Leukemia Viral Oncogene Homolog 3 using the numbering given in entry NP_001973.2 (GeneID 2065).
0.1 mL
Primary
2065
Western Blot
Western Blot
c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Rabbit
IgG
Store at 4°C in the dark.
Polyclonal
Human