Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ErbB3 Recombinant Rabbit Monoclonal Antibody (138), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

$221.00 - $430.00

Specifications

Antigen ErbB3
Clone 138
Concentration 1 mg/mL
Applications ELISA
Classification Recombinant Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIMA543092
Encompass
View Documents
Thermo Scientific
MA543092
100 μL
Each for $430.00
Add to cart
 
PIMA543528
Encompass
View Documents
Thermo Scientific
MA543528
50 μL
Each for $221.00
Add to cart
 
Description

Description

Specificity: Mouse HER3/ErbB3

ErbB3 Recombinant Monoclonal antibody specifically detects ErbB3 in Mouse samples. It is validated for ELISA
Specifications

Specifications

ErbB3
1 mg/mL
Recombinant Monoclonal
Liquid
RUO
PBS with no preservative
avian erythroblastosis oncogene B 3; avian erythroblastosis oncogene B 3 receptor; C76256; c-erbB3; c-erbB-3; C-errB-3; erb-b2 receptor tyrosine kinase 3; ERBB3; Erbb-3; Erbb3r; erbB3-S; glial growth factor receptor; HER3; human epidermal growth factor receptor 3; LCCS2; MDA BF 1; MDA-BF-1; MGC88033; nuc-ErbB3; p180 ErbB3; p180-ErbB3; p45 sErbB3; p45-sErbB3; p85 sErbB3; p85-sErbB3; proto-oncogene-like protein c-ErbB-3; receptor tyrosine kinase; receptor tyrosine-protein kinase erbB-3; receptor tyrosine-protein kinase erbB-3; LOW QUALITY PROTEIN: receptor tyrosine-protein kinase erbB-3; tyrosine kinase-type cell surface receptor HER3; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
ERBB3
IgG
Protein A
Antibody
138
ELISA
Unconjugated
Rabbit
Mouse
Q61526
13867
Recombinant Mouse HER3/ErbB3 protein.
Primary
-20° C, Avoid Freeze/Thaw Cycles
ERBB3
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit