Sino Biological™ ERBB3 Recombinant Human Protein (without Catalytic Activity Domain), His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding extracellular domain (Met 1-Thr 643) of human ERBB3 precursor fused to polyhistidine tag

Manufacturer: Sino Biological™ 10201H08H250

 View more versions of this product

Catalog No. 10-201-H08H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Product Type ERBB3 Recombinant Human Protein
    Molecular Weight 70.2kd
    Purity ≥85% by SDS-PAGE
    Quantity 5 x 50μg
    Source Human
    Storage Requirements −20° to −70°C
    Structural Form Truncated, Cell Surface Protein, Recombinant
    Endotoxin Concentration <1 EU/μg
    Common Name V-erb-b2 erythroblastic leukemia viral oncogene homolog 3
    Gene Symbol ERBB3
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Growth Factors and Receptors
    Expression System Mammalian
    For Use With (Application) Studying protein-protein interactions and protein structure analysis
    Format Lyophilized
    Gene ID (Entrez) 2065

    Expressed from DNA sequence encoding extracellular domain (Met 1-Thr 643) of human ERBB3 precursor fused to polyhistidine tag

    • Activity is measured by its binding ability in functional ELISA
    • Immobilized human ERBB3-Fc at 1µg/well can bind biotinylated recombinant human NRG1 w/linear range of 0.032-0.16µg/mL
    • ED50 of human NRG1 is 0.43 µg/ml

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit