Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ErbB4/Her4 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB11312
This item is not returnable.
View return policy
Description
ErbB4/Her4 Monoclonal specifically detects ErbB4/Her4 in Human samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
ErbB4/Her4 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian) | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2066 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Blockade of Receptor-ligand Interaction | |
918903 | |
Blockade of Receptor-ligand Interaction 0.2-1 ug/mL | |
Q15303 | |
ERBB4 | |
Mouse myeloma cell line NS0-derived recombinant human ErbB4/Her4 Gln26-Arg649 Accession # Q15303 | |
100 μg | |
Primary | |
Detects human ErbB4/Her4 in direct ELISAs. | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction