Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ErbB4/Her4 Mouse anti-Human, Clone: 918903, R&D Systems™

Mouse Monoclonal Antibody

$128.24 - $367.39

Specifications

Antigen ErbB4/Her4
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Format Purified
Clone 918903
Applications Blockade of Receptor-ligand Interaction
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB11312
View Documents
R&D Systems
MAB11312
100 ug
Each for $367.39
Add to cart
 
MAB11312SP
View Documents
R&D Systems
MAB11312SP
25 ug
Each for $128.24
Add to cart
 
Description

Description

ErbB4/Her4 Monoclonal specifically detects ErbB4/Her4 in Human samples. It is validated for Blockade of Receptor-ligand Interaction.
Specifications

Specifications

ErbB4/Her4
Purified
Blockade of Receptor-ligand Interaction
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
918903
Unconjugated
IgG2b
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit