Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ErbB4/Her4 Mouse anti-Human, Mouse, Clone: HFR-1, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen ErbB4/Her4
Clone HFR-1
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Flow Cytometry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121793
View Documents
Novus Biologicals
NBP30716010UG
100 μg
Each for $352.89
Add to cart
 
NB121794
View Documents
Novus Biologicals
NBP30716020UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

ErbB4/Her4 Monoclonal antibody specifically detects ErbB4/Her4 in Human, Mouse samples. It is validated for Flow Cytometry, Immunofluorescence
Specifications

Specifications

ErbB4/Her4
0.2 mg/ml
Flow Cytometry, Immunofluorescence
Purified
RUO
Human, Mouse
avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian)
Recombinant human ErbB4/Her4 protein fragment (around aa 1116-1269) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
HFR-1
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Unconjugated
Mouse
Angiogenesis, Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors
10 mM PBS with 0.05% BSA
2066
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit