Learn More
ERbeta/NR3A2 Mouse anti-Human, Janelia Fluor 646, Clone: 4D2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP141193JF646
Catalog No. NBP14193646
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
ERbeta/NR3A2Monoclonalantibody specifically detects ERbeta/NR3A2 in Human samples.
Specifications
Specifications
ERbeta/NR3A2
4D2
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
ESR2
Amino acids 1-153 of human ER expressed in E. coli.
Protein G purified
Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Dendritic Cell Markers, GPCR, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Primary
2100
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Erb, ER-beta, ESR-BETA, ESTRB, estrogen receptor 2 (ER beta), estrogen receptor beta, estrogen receptor beta 4, NR3A2ESRB, Nuclear receptor subfamily 3 group A member 2
Mouse
IgG2b
0.1mL
Store at 4°C in the dark.
Monoclonal
This antibody will recognize human ER beta.