Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Erbium foil, 0.1mm (0.004 in.) thick, 99.9% (REO)

Click to view available options
Quantity:
50 x 50 mm
7440-52-0
Er
167.259
MFCD00010987
UYAHIZSMUZPPFV-UHFFFAOYSA-N
unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic
23980
CHEBI:33379
erbium
[Er]
7440-52-0
Er
167.259
MFCD00010987
UYAHIZSMUZPPFV-UHFFFAOYSA-N
unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic
23980
CHEBI:33379
erbium
[Er]
Quantity 50 x 50 mm
Weight ≈0.57g/25x25mm
UN Number UN3178
Solubility Information Soluble in acids. Insoluble in water.
Physical Form Foil
Percent Purity 99.9%
Color Silver to Gray
Chemical Name or Material Erbium foil, 0.1mm (0.004 in.) thick
Assay (REO)

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-160-1

TSCA : Yes

Recommended Storage : Ambient temperatures