Alfa Aesar™ Erbium(III) acetate tetrahydrate, 99% (REO)

Manufacturer: Alfa Aesar™ 4589714

Quantity 25g

 View more versions of this product

Catalog No. AA4589714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15280-57-6
    Molecular Formula C6H9ErO6·4H2O
    Linear Formula Er(OOCCH3)3·4H2O
    Formula Weight 416.45 (344.39 Anhydrous)
    MDL Number MFCD00013034
    Chemical Name or Material Erbium (III) acetate tetrahydrate

    Specifications

    Formula Weight 416.45 (344.39 Anhydrous)
    Physical Form Crystalline Aggregates
    Odor Odorless
    MDL Number MFCD00013034
    Color Pink
    TSCA TSCA
    Molecular Formula C6H9ErO6·4H2O
    Linear Formula Er(OOCCH3)3·4H2O
    CAS 15280-57-6
    Density 2.114g/mL
    Sensitivity Hygroscopic
    Quantity 25g
    Chemical Name or Material Erbium (III) acetate tetrahydrate
    Assay 99%, (REO)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit