Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Erbium(III) chloride, ultra dry, 99.9% (REO)

Click to view available options
Quantity:
5g
25g
10138-41-7
Cl3Er
273.609
MFCD00010989
HDGGAKOVUDZYES-UHFFFAOYSA-K
erbium chloride,erbium trichloride,erbium chloride ercl3,ercl3,erbium iii chloride, anhydrous,erbium iii chloride, ultra dry,erbium iii chloride hexahydrate,erbium iii chloride, anhydrous, ?ercl3,erbium iii chloride, anhydrous, powder trace metals basis,erbium iii chloride, anhydrous, beads,-10 mesh trace metals basis
66277
trichloroerbium
Cl[Er](Cl)Cl
10138-41-7
Cl3Er
273.609
MFCD00010989
HDGGAKOVUDZYES-UHFFFAOYSA-K
erbium chloride,erbium trichloride,erbium chloride ercl3,ercl3,erbium iii chloride, anhydrous,erbium iii chloride, ultra dry,erbium iii chloride hexahydrate,erbium iii chloride, anhydrous, ?ercl3,erbium iii chloride, anhydrous, powder trace metals basis,erbium iii chloride, anhydrous, beads,-10 mesh trace metals basis
66277
trichloroerbium
Cl[Er](Cl)Cl
  • Ampouled under argon
Solubility Information Soluble in water,strong mineral acids. Slightly soluble in MeOH and THF.
Formula Weight 273.62
Physical Form Powder
Percent Purity 99.9%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Boiling Point 1500°C
Melting Point 774°C
Quantity 25g
Chemical Name or Material Erbium(III) chloride, Ultra dry
Assay (REO)
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 233-385-0

RTECSNumber : KD8575000

TSCA : Yes

storageNote1 :

  • Ampouled under argon

Recommended Storage : Ambient temperatures