Alfa Aesar™ Erbium(III) iodide, anhydrous, 90% min, (REO)

Manufacturer: Alfa Aesar™ 3354009

Quantity 10g

 View more versions of this product

Catalog No. AA3354009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13813-42-8
    Formula Weight 547.97
    MDL Number MFCD00016075
    Chemical Name or Material Erbium(III) iodide, Anhydrous

    Specifications

    Formula Weight 547.97
    Boiling Point 1280°C
    Physical Form Crystalline
    MDL Number MFCD00016075
    TSCA TSCA
    Sensitivity Hygroscopic
    CAS 13813-42-8
    Melting Point 1020°C
    Quantity 10g
    Chemical Name or Material Erbium(III) iodide, Anhydrous
    Assay ≤90%, (REO)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit