Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erbium(III) oxide, 99.5% (REO), Alfa Aesar™

S. A. 13 to 17m2/g

Manufacturer:  Alfa Aesar 4466818

Quantity 50g

 View more versions of this product

Catalog No. AA4466818


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12061-16-4
99.5% (REO)
382.56
Erbia
Powder or Lump
Er2O3
MFCD00010991
Erbium(III) oxide

Specifications

12061-16-4
99.5% (REO)
Powder or lump
MFCD00010991
50g
Powder or Lump
Er2O3
382.56
Erbia
Erbium(III) oxide

Safety and Handling

SDS & Safety Documents

H319: Causes serious eye irritation.

EINECSNumber : 235-045-7

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit