Erbium(III) oxide, 99.5% (REO), Alfa Aesar™

S. A. 13 to 17m2/g

Manufacturer: Alfa Aesar 4466809

Quantity 10g

 View more versions of this product

Catalog No. AA4466809

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 12061-16-4
    Physical Form Powder or Lump
    Assay Percent Range 99.5% (REO)
    Molecular Formula Er2O3
    Formula Weight 382.56
    MDL Number MFCD00010991
    Synonym Erbia
    Chemical Name or Material Erbium(III) oxide

    Specifications

    Physical Form Powder or Lump
    Formula Weight 382.56
    Quantity 10g
    Density 8.64 g/cm3
    EINECS Number 235-045-7
    MDL Number MFCD00010991
    Assay Percent Range 99.5% (REO)
    Molecular Formula Er2O3
    Appearance Powder or lump
    CAS 12061-16-4
    Synonym Erbia
    Health Hazard 1 H319: Causes serious eye irritation.
    Chemical Name or Material Erbium(III) oxide

    H319: Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit