Alfa Aesar™ Erbium(III) oxide, REacton™, 99% (REO)

Manufacturer: Alfa Aesar™ 66222A1

Quantity 1kg

 View more versions of this product

Catalog No. AA66222A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12061-16-4
    Formula Weight 382.56
    MDL Number MFCD00010991
    Merck Index 14,3646
    Chemical Name or Material Erbium(III) oxide
    Grade Reacton™

    Specifications

    Formula Weight 382.56
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    Sensitivity Hygroscopic
    MDL Number MFCD00010991
    CAS 12061-16-4
    Melting Point 2400°C
    Health Hazard 1 Warning
    Odor Odorless
    Quantity 1kg
    Density 8.64g/mL
    RTECS Number KD9250000
    TSCA TSCA
    Merck Index 14,3646
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Erbium(III) oxide
    Assay 99%, (REO)
    Grade Reacton™

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit