Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erbium Pieces, 99.9% (REO), Alfa Aesar™

Click to view available options
Quantity:
1g
10g
50g
Packaging:
Glass Bottle under Argon
Packaged under argon in glass bottle
7440-52-0
Er
167.259
MFCD00010987
UYAHIZSMUZPPFV-UHFFFAOYSA-N
unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic
23980
CHEBI:33379
erbium
[Er]
7440-52-0
Er
167.259
MFCD00010987
UYAHIZSMUZPPFV-UHFFFAOYSA-N
unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic
23980
CHEBI:33379
erbium
[Er]
Assay Percent Range 99.9% (REO)
Appearance Silver Gray Pieces
Physical Form Pieces
Density 9.066g/cm3
Impurities Total Rare Earth Oxide Impurities: 0.1% max.
Packaging Packaged under argon in glass bottle
Color Silver to Gray
Quantity 1g
Chemical Name or Material Erbium, Rare Earth Oxides Content

Keep container tightly closed and dry.

H228-H260; Flammable solid. In contact with water releases flammable gases which may ignite spontaneously.

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-154-9