Learn More
Alfa Aesar™ Erbstatin Analog
An inhibitor of the EGF receptor associated tyrosine kinase
Manufacturer:  Alfa Aesar™ J61612MCR
 View more versions of this product
Catalog No. AAJ61612MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1mg
−30° to −10°C
An inhibitor of the EGF receptor associated tyrosine kinase
C10H10O4
Methyl 2,5-dihydroxycinnamate
Signal Transduction Reagent
Yellow
Powder
63177-57-1
194.19g/mol
Soluble in methanol or 0.1M NaOH; Insoluble in water