Learn More
ERCC1 Antibody (OTI1A3), Biotin, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270662B
Catalog No. NBP270662B
Add to Cart
Description
Description
ERCC1 Monoclonal specifically detects ERCC1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
ERCC1
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
ERCC1
Full-length protein expressed in 293T cell transfected with human ERCC1 expression vector.
0.1 mL
Primary
Human, Mouse
IgG1
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI1A3
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20
Mouse
Affinity Purified
Cancer, DNA Repair, Nucleotide Excision Repair
2067.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions