Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERCC1 Mouse anti-Human, Clone: OTI6H7, lyophilized, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene CF805691
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The mammalian ERCC1 (Excision Repair Cross Complementing) polypeptide is required for nucleotide excision repair (NER) of damaged DNA and is homologous to Saccharomyces cerevisiae RAD10, which functions in repair and mitotic intrachromosomal recombination. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells XPG cleaves 3 ft. of the DNA lesion while the ERCC1-XPF complex makes the 5 ft. incision.Specifications
ERCC1 | |
Monoclonal | |
Unconjugated | |
ERCC1 | |
Lyophilized | |
ERCC1 | |
Full length human recombit protein of human ERCC1 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Immunohistochemistry (Paraffin), Western Blot | |
OTI6H7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P07992 | |
COFS4, RAD10, UV20 | |
Mouse | |
IgG2a | |
100 μg | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
2067 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title