Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC1 Mouse anti-Human, Clone: ERCC1/2683, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen ERCC1
Clone ERCC1/2683
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA, Protein Array
Applications Western Blot, ELISA, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121798
View Documents
Novus Biologicals
NBP30716220UG
20 μg
Each for $138.37
Add to cart
 
NB121797
View Documents
Novus Biologicals
NBP30716210UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

ERCC1 Monoclonal antibody specifically detects ERCC1 in Human samples. It is validated for Western Blot, ELISA, Protein Array
Specifications

Specifications

ERCC1
0.2 mg/ml
Western Blot, ELISA, Peptide Array
Purified
RUO
Human
COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20
Recombinant fragment (around aa 191-281) of human ERCC1 protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
ERCC1/2683
Western Blot 1-2 ug/ml, ELISA, Protein Array
Unconjugated
Mouse
Cancer, DNA Repair, Nucleotide Excision Repair
10 mM PBS with 0.05% BSA
2067
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit