Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC1 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI2H1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270685JF549

Catalog No. N270685J549


Add to cart

Description

Description

ERCC1 Monoclonal antibody specifically detects ERCC1 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

ERCC1
OTI2H1
Immunohistochemistry
Affinity Purified, Conjugated
ERCC1
Full length recombinant protein of human ERCC1 (NP_001974) produced in HEK293T cells.
Immunogen affinity purified
Cancer, DNA Repair, Nucleotide Excision Repair
Primary
2067.0
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
COFS4, DNA excision repair protein ERCC-1, excision repair cross-complementing rodent repair deficiency, complementationgroup 1 (includes overlapping antisense sequence), RAD10, UV20
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit