Learn More
Invitrogen™ ERCC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109691
Description
Immunogen sequence: CPMSPEFYRE YVAIKTKKRI LLYTMNPNKF RACQFLIKFH ERRNDKIIVF ADNVFALKEY AIRLNKPYIY GPTSQGERMQ ILQNFKHNPK INTIFISKVG DTSFDLPEAN VLIQISSHGG SRRQEAQRLG RVLRAKKGMV AEEYNAFFYS LVSQDTQEMA YSTKRQRFLV DQGYSFKVIT KLAGMEEEDL AFSTKEEQQQ LLQKVLAATD LDAEEEVVAG EFGSRSSQAS RRFGTMSSMS GADDTVYMEY HSSRSKAPSK HVHPLFKRFR K.
The human XPB DNA helicase is a subunit of the DNA repair/basal transcription factor TFIIH, and is involved in early steps of the nucleotide excision repair (NER) pathway. Two distinct clinical phenotypes, xeroderma pigmentosum associated with Cockayne's syndrome (XP/CS) and trichothiodystrophy (TTD), can be due to mutations in the XPB gene.
Specifications
ERCC3 | |
Polyclonal | |
Unconjugated | |
Ercc3 | |
Basic transcription factor 2 89 kDa subunit; BTF2; BTF2 p89; BTF2-p89; DNA excision repair protein ERCC-3; DNA repair helicase; DNA repair protein complementing XP-B cells; ERCC excision repair 3, TFIIH core complex helicase subunit; Ercc3; Ercc-3; excision repair 3; excision repair cross-complementation group 3; excision repair cross-complementing rodent repair deficiency, complementation group 3; General transcription and DNA repair factor IIH helicase subunit XPB; GTF2H; RAD25; TFIIH; TFIIH 89 kDa subunit; TFIIH basal transcription factor complex 89 kDa subunit; TFIIH basal transcription factor complex helicase XPB subunit; TFIIH p89; TFIIH subunit XPB; TTD2; Xeroderma pigmentosum group B-complementing protein; xeroderma pigmentosum, complementation group B; XPB; XPBC | |
Rabbit | |
Affinity Chromatography | |
RUO | |
13872, 2071, 291703 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P19447, P49135, Q4G005 | |
Ercc3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 512-782 of human ERCC3 (NP_000113.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.