Learn More
ERCC6/CSB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 205481AP
Catalog No. 50-173-2953
Add to cart
Description
Description
The antibody is specific to ERCC6.
This gene encodes a member of the GTP cyclohydrolase family. The encoded protein is the first and rate-limiting enzyme in tetrahydrobiopterin biosynthesis, catalyzing the conversion of GTP into 7,8-dihydroneopterin triphosphate. BH4 is an essential cofactor required by aromatic amino acid hydroxylases as well as nitric oxide synthases. Mutations in this gene are associated with maligt hyperphenylalaninemia and dopa-responsive dystonia. Several alternatively spliced transcript variants encoding different isoforms have been described; however, not all variants give rise to a functional enzyme.
Specifications
Specifications
ERCC6/CSB
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q03468
ARMD5, ATP dependent helicase ERCC6, CKN2, Cockayne syndrome protein CSB, COFS, COFS1, CSB, ERCC6, RAD26
Rabbit
IgG
150 μL
-20°C
Primary
2074, 306274, 319955
Western Blot
Unconjugated
ERCC6
Liquid
Ercc6
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat