Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Abnova™ ERCC8 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00001161T02
Catalog No. 89-943-559
Specifications
Specifications
Western Blot
293T
NM_000082.2
pCMV-ERCC8 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
ERCC8 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 43.67kDa
Transient overexpression cell lysate was tested with anti-ERCC8 antibody (Mfr. No. H00001161-B02) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)