Learn More
Aviva Systems Biology Corp ERCC8 Antibody - N-terminal region (ARP57955_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP57955_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERCC8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ERCC8. Alternate names: CKN1, CSAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62190