Learn More
Aviva Systems Biology Corp ERCC8 Antibody - middle region (ARP58570_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58570_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERCC8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ERCC8. Alternate names: CKN1, CSAApplicationWB. Species reactivity: Human
Catalog No. 50-014-62357