Learn More
ERCC8 Antibody (OTI2G1), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270412IR
Catalog No. NBP270412IR
Add to Cart
Description
Description
ERCC8 Monoclonal specifically detects ERCC8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
ERCC8
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
ERCC8
Full length recombinant protein of human ERCC8 (NP_001007235) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG2a
Western Blot, Immunohistochemistry
OTI2G1
Western Blot, Immunohistochemistry
CKN1DNA excision repair protein ERCC-8, Cockayne syndrome 1 (classical), Cockayne syndrome WD repeat protein CSA, CSACockayne syndrome WD-repeat protein CSA, excision repair cross-complementing rodent repair deficiency, complementationgroup 8
Mouse
Affinity Purified
Cancer, DNA Repair, Nucleotide Excision Repair
1161.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions