Learn More
ERCC8 Antibody (OTI2G1), FITC, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270412F
Catalog No. NBP270412F
Add to Cart
Description
Description
ERCC8 Monoclonal specifically detects ERCC8 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
Specifications
ERCC8
Monoclonal
FITC
PBS with 0.05% Sodium Azide
ERCC8
Full length recombinant protein of human ERCC8 (NP_001007235) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG2a
Immunohistochemistry
OTI2G1
Immunohistochemistry
CKN1DNA excision repair protein ERCC-8, Cockayne syndrome 1 (classical), Cockayne syndrome WD repeat protein CSA, CSACockayne syndrome WD-repeat protein CSA, excision repair cross-complementing rodent repair deficiency, complementationgroup 8
Mouse
Affinity Purified
Cancer, DNA Repair, Nucleotide Excision Repair
1161.0
Store at 4°C in the dark.
Recommendations
Recommendations