Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC8 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI2G1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270412JF549

Catalog No. N270412J549


Add to cart

Description

Description

ERCC8 Monoclonal antibody specifically detects ERCC8 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

ERCC8
OTI2G1
Immunohistochemistry
Affinity Purified, Conjugated
ERCC8
Full length recombinant protein of human ERCC8 (NP_001007235) produced in E.coli.
Immunogen affinity purified
Cancer, DNA Repair, Nucleotide Excision Repair
Primary
1161.0
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CKN1DNA excision repair protein ERCC-8, Cockayne syndrome 1 (classical), Cockayne syndrome WD repeat protein CSA, CSACockayne syndrome WD-repeat protein CSA, excision repair cross-complementing rodent repair deficiency, complementationgroup 8
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit