Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eRF1 Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI2E3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270692JF646

Catalog No. N270692J646


Add to cart

Description

Description

eRF1 Monoclonal antibody specifically detects eRF1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

eRF1
OTI2E3
Flow Cytometry
Affinity Purified, Conjugated
ETF1
Full length recombinant protein of human eRF1 (NP_004721) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
D5S1995, ERF1eRF1, eukaryotic peptide chain release factor subunit 1, Eukaryotic release factor 1, eukaryotic translation termination factor 1, polypeptide chain release factor 1, Protein Cl1, RF1ERF, sup45 (yeast omnipotent suppressor 45) homolog-like 1, SUP45L1, TB3-1MGC111066
Mouse
IgG2a
0.1 mL
Primary
2107.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit