Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERG Mouse anti-Human, Clone: ERG/2107, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30717220UG
Description
ERG Monoclonal specifically detects ERG in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
ERG | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
transcriptional regulator ERG, 10ets-related, erg-3, p55, TMPRSS2-ERG prostate cancer specific, transcriptional regulator ERG, Transforming protein ERG, v-ets erythroblastosis virus E26 oncogene homolog (avian), v-ets erythroblastosis virus E26 oncogene like, v-ets erythroblastosis virus E26 oncogene like (avian) | |
Recombinant human full-length ERG protein | |
20 μg | |
Cancer | |
2078 | |
Store at 4°C. | |
IgG2b κ |
Immunohistochemistry (Paraffin), Peptide Array | |
ERG/2107 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction