Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERG Rabbit anti-Human, Clone: ERG/22R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen ERG
Clone ERG/22R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121820
View Documents
Novus Biologicals
NBP30717320UG
20 μg
Each for $159.82
Add to cart
 
NB121819
View Documents
Novus Biologicals
NBP30717310UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

ERG Monoclonal antibody specifically detects ERG in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

ERG
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
transcriptional regulator ERG, 10ets-related, erg-3, p55, TMPRSS2-ERG prostate cancer specific, transcriptional regulator ERG, Transforming protein ERG, v-ets erythroblastosis virus E26 oncogene homolog (avian), v-ets erythroblastosis virus E26 oncogene like, v-ets erythroblastosis virus E26 oncogene like (avian)
Synthetic peptide corresponding to residues within aa450 to C-terminus of ERG was used as an immunogen.
Monoclonal
Protein A or G purified
ERG/22R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Cancer
10 mM PBS with 0.05% BSA
2078
IgG Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit