Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERG (TMPRSS2-ERG) Rabbit anti-Human, Polyclonal, Biologo Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$821.59 - $1599.62

Specifications

Antigen ERG (TMPRSS2-ERG)
Clone EP111
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0758 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
ERG00205
0.5 mL
Each for $821.59
Add to cart
 
50-210-0759 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
ERG00210
1 mL
Each for $1,599.62
Add to cart
 
Description

Description

ERG (TMPRSS2-ERG) Polyclonal antibody specifically detects ERG (TMPRSS2-ERG) in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

ERG (TMPRSS2-ERG)
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
P11308
2078
IgG
Primary
4° C
Erg
EP111
Polyclonal
Liquid
RUO
PBS with 0.2% BSA and 15mM sodium azide; pH
avian erythroblastosis virus E-26 (v-ets) oncogene related; D030036I24Rik; Erg; ERG, ETS transcription factor; Erg-3; erythroblast transformation-specific transcription factor ERG variant 10; ets-related; H Kv11.3; HGNC:3446; OTTHUMP00000115970; OTTHUMP00000208036; p55; TMPRSS2/ERG fusion; transcriptional regulator ERG; transcriptional regulator ERG (transforming protein ERG); Transforming protein ERG; v-ets avian erythroblastosis virus E26 oncogene; v-ets avian erythroblastosis virus E26 oncogene homolog; v-ets avian erythroblastosis virus E26 oncogene related; v-ets erythroblastosis virus E26 oncogene homolog; v-ets erythroblastosis virus E26 oncogene like
Purified human myeloma-IgG polyaminostyrene conjugate
Polyclonal
Protein A/G
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit