Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERGIC2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548793

Catalog No. PA548793

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

Possible role in transport between endoplasmic reticulum and Golgi.
Specifications

Specifications

ERGIC2
Polyclonal
PBS with 0.09% sodium azide
Q96RQ1
Endoplasmic reticulum-Golgi intermediate compartment protein 2, ERGIC2, ERV41, PTX1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51290
Western Blot
Unconjugated
ERGIC2
Liquid
ERGIC2
KLH conjugated synthetic peptide between 335-362 amino acids from the C-terminal region of human ERGIC2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit