Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ERGIC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531996
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (96%), Rat (97%), Zebrafish (86%), Xenopus laevis (83%), Rabbit (97%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ERGIC3 is possible role in transport between endoplasmic reticulum and Golgi.
Specifications
ERGIC3 | |
Polyclonal | |
Unconjugated | |
ERGIC3 | |
2310015B14Rik; AV318804; C20orf47; CGI-54; D2Ucla1; dJ477O4.2; Endoplasmic reticulum-Golgi intermediate compartment protein 3; endoplasmic reticulum-localized protein ERp43; ERGIC and golgi 3; ERGIC3; ERV46; NY-BR-84; PRO0989; Sdbcag84; serologically defined breast cancer antigen 84; serologically defined breast cancer antigen NY-BR-84; Serologically defined breast cancer antigen NY-BR-84 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51614 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.65 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y282 | |
ERGIC3 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 383 of Human ERGIC3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction