Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ERGIC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553401
Description
Immunogen sequence: NINMTHYIQH LSFGEDYPGI VNPLDHTNVT APQASMMFQY FVKVVPTVYM KVDGEVLRTN QFSVTRHEKV ANGLLGDQGL PGVFVLYELS PMMVKLTEKH Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
ERGIC3 is possible role in transport between endoplasmic reticulum and Golgi.
Specifications
ERGIC3 | |
Polyclonal | |
Unconjugated | |
ERGIC3 | |
2310015B14Rik; AV318804; C20orf47; CGI-54; D2Ucla1; dJ477O4.2; Endoplasmic reticulum-Golgi intermediate compartment protein 3; endoplasmic reticulum-localized protein ERp43; ERGIC and golgi 3; ERGIC3; ERV46; NY-BR-84; PRO0989; Sdbcag84; serologically defined breast cancer antigen 84; serologically defined breast cancer antigen NY-BR-84; Serologically defined breast cancer antigen NY-BR-84 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51614 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y282 | |
ERGIC3 | |
Recombinant protein corresponding to Human ERGIC3. Recombinant protein control fragment (Product #RP-90950). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction