Learn More
Fisherbrand™ Ergo-Fit™ Pipette
GSA_VA
Ergo-Fit contoured shape pipet with adjustable hook fits hand like a glove
$940.00 - $1233.00
Specifications
Accuracy Multiple
Autoclavable Autoclavable
Number of Channels 1
Increments Multiple
Precision Multiple
View More Specs
Products 2
Catalog Number Volume (Metric) Price Quantity & Availability  
Catalog Number Volume (Metric) Price Quantity & Availability  
12-146-681
GSA_VA
View Documents
0.2-2, 2-20, 20-200, 100-1000 µL
Each for $1,233.00
Add to cart
12-146-682
GSA_VA
View Documents
1-10, 10-100, 100-1000 µL
Each for $940.00
Add to cart
Description
Description
  • Low force plunger enables light thumb action pipet for liquid handling
  • Plunger helps to manually control speed of aspirating and dispensing with minimal effort
  • Audible click for gentle tip acquisition and push button for tip discard
  • Cap opener at base for easy opening of PCR and microtubes
  • Simple in-lab calibration
  • Ergonomically designed pipet ensures pipetting comfort
  • Plunger allows to adjust volume for fast pipetting
  • Available in six models from 0.2 to 1000μL
  • Autoclavable pipet
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Multiple
1
Multiple
Autoclavable
Multiple
Two-year Warranty