Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergocalciferol, 97.0 to 103.0%, Affymetrix/USB™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Plastic bottle
50-14-6
C28H44O
396.659
MFCD00166988
MECHNRXZTMCUDQ-FWJFVZHNSA-N
ergocalciferol,vitamin d2,calciferol,viosterol,oleovitamin d2,ercalciol,ergorone,detalup,condocaps,crystallina
134129658
(1S)-3-[2-[(1S,7aR)-1-[(2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
CC(C)C(C)C=CC(C)C1CCC2C1(CCCC2=CC=C3CC(CCC3=C)O)C
50-14-6
C28H44O
396.659
MFCD00166988
MECHNRXZTMCUDQ-FWJFVZHNSA-N
ergocalciferol,vitamin d2,calciferol,viosterol,oleovitamin d2,ercalciol,ergorone,detalup,condocaps,crystallina
134129658
(1S)-3-[2-[(1S,7aR)-1-[(2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
CC(C)C(C)C=CC(C)C1CCC2C1(CCCC2=CC=C3CC(CCC3=C)O)C
Quantity 100g
Melting Point 118°C to 119°C
Packaging Plastic bottle
UN Number UN2811
Formula Weight 396.65
Physical Form Powder
Percent Purity 97.0 to 103.0%
Chemical Name or Material Ergocalciferol