Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergocalciferol, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
10 g
100 g
1 kg
Packaging:
Amber Glass Bottle
50-14-6
C28H44O
396.66
MECHNRXZTMCUDQ-VLOQVYPSSA-N
(1S,3E)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol
CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C
50-14-6
C28H44O
396.66
MECHNRXZTMCUDQ-VLOQVYPSSA-N
(1S,3E)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol
CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C

ER100, 50-14-6, C28H44O

Spectrum™ Ergocalciferol, USP or Vitamin D2 is used with other medications to treat low levels of calcium or phosphate. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Melting Range 115-119°C
Specific Rotation +103° to +106°
Quantity 10 g
CAS Max % 1
UN Number UN2811
Percent Purity 97 to 103%